missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC93 Antibody (OTI6E11), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71963JF549
This item is not returnable.
View return policy
Description
CCDC93 Monoclonal specifically detects CCDC93 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CCDC93 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CCDC93 | |
| Recombinant protein fragment corresponding to amino acids 293-631 of human CCDC93 (NP_061917) produced in E.coli. | |
| 0.1 mL | |
| 54520.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI6E11 | |
| Western Blot, Immunohistochemistry | |
| coiled-coil domain containing 93, coiled-coil domain-containing protein 93, FLJ10996, FLJ25197, MGC13033 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction