Learn More
CCDC96 Polyclonal Antibody
GREENER_CHOICE
CCDC96 Polyclonal Antibody
Brand:  Invitrogen PA570126
Product Code. 15868533
  • € 530.00 / 100µL
Description
CCDC96 Polyclonal Antibody for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCDC96
Polyclonal
Unconjugated
CCDC96
4921513E08Rik; CCDC96; coiled-coil domain containing 96; coiled-coil domain-containing protein 96; RGD1305153
Rabbit
Affinity Chromatography
RUO
257236
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q2M329
CCDC96
synthetic peptide directed towards the N terminal region of human CCDC96.