missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC99 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-61595CL1
This item is not returnable.
View return policy
Description
CCDC99 Polyclonal antibody specifically detects CCDC99 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| CCDC99 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 54908 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| Arsenite-related gene 1 protein, coiled-coil domain containing 99, Coiled-coil domain-containing protein 99, FLJ20364, FLJ40690, hSpindly, protein Spindly, Rhabdomyosarcoma antigen MU-RMS-40.4A, rrhabdomyosarcoma antigen protein MU-RMS-40.4A, Spindly | |
| The immunogen recognized by this antibody maps to a region between residue 525 and 575 of human SPINDLY (coiled-coil domain containing 99) using the numbering given in entry NP_060255.2 (GeneID 54908). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction