missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Motic™ CCIS™ LWD Plan Achromatic Objective
Motic™ CCIS™ LWD Plan achromatic objective is compatible with all AE inverted microscopes
€ 220.00 - € 297.00
Specifications
Type | Plan |
---|---|
For Use With (Equipment) | AE inverted microscopes |
Quantity | 1 Piece |
Product Code | Brand | Magnification Power | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Magnification Power | Price | Quantity & Availability | |||||
11732348
|
Motic™
1101001703251 |
20X |
€ 220.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11742348
|
Motic™
1101001703261 |
40X |
€ 297.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
Description
- For use with AE31 Inverted Microscope
- Long Working Distance
- Achromatic Plan Objective
- Flat field
- Eliminates color aberration
- Magnification and Numerical Apertures available:
- 20X/0.3 (WD=4.7mm)
- 40X/0.5 (WD=3.0mm)
Specifications
Plan | |
AE inverted microscopes | |
1 Piece |