missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCK-AR Antibody (377251), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2680K
This item is not returnable.
View return policy
Description
CCK-AR Monoclonal antibody specifically detects CCK-AR in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| CCK-AR | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 377251 | |
| Flow Cytometry, CyTOF-ready | |
| CCK1-R, CCK-A, CCK-A receptor, CCK-AR, CCKRA, cholecystokinin A receptor, cholecystokinin receptor type A, cholecystokinin type-A receptor, Cholecystokinin-1 receptor | |
| NS0 mouse myeloma cell line transfected with human CCK-A R, Met1-Gln428, Accession # P32238 | |
| 0.1 mL | |
| Diabetes Research, GPCR, Lipid and Metabolism | |
| 886 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction