Learn More
Invitrogen™ CCK Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA137267
 View more versions of this product
Product Code. 11501693
  • € 808.00 / 1mL
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is stomach tissue.
Cholecystokinin (CCK) is a neuropeptide hormone and neurotransmitter widely distributed throughout the gastrointestinal (GI) tract and the central nervous system (CNS). Antibodies that react specifically with CCK are useful for the study of the mode of action, differential tissue expression and intracellular and subcellular localization of CCK in the CNS and periphery as well as in neuroendocrine cells of the digestive system.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCK
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.6
P06307
CCK
Synthetic peptide within Human Cholecystokinin aa 50 to the C-terminus (N terminal).
1 mL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
CCK
(1-49)-CCK58; CCK; CCK12; CCK18; CCK22; CCK25; CCK33; CCK39; CCK5; CCK58; CCK7; CCK8; Cholecystokinin; cholecystokinin octapeptide; cholecystokinin triacontatriapeptide; Cholecystokinin-12; Cholecystokinin-18; Cholecystokinin-22; Cholecystokinin-25; Cholecystokinin-33; Cholecystokinin-39; Cholecystokinin-5; Cholecystokinin-58; Cholecystokinin-58 desnonopeptide; Cholecystokinin-7; Cholecystokinin-8; prepro-cholecystokinin
Rabbit
Antigen affinity chromatography
RUO
885
-20°C, Avoid Freeze/Thaw Cycles