Learn More
CCL1/I-309/TCA-3 Antibody (RM0167-6H50), FITC, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NBP2-11876F
Product Code. 18460702
  • € 607.00 / 0.10mL
Description
CCL1/I-309/TCA-3 Monoclonal antibody specifically detects CCL1/I-309/TCA-3 in Mouse samples. It is validated for Western Blot, Neutralization
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCL1/I-309/TCA-3
Monoclonal
FITC
PBS
Rat
Protein A or G purified
RUO
Primary
Mouse
Purified
Western Blot, Neutralization
RM0167-6H50
Western Blot, Block/Neutralize
C-C motif chemokine 1, chemokine (C-C motif) ligand 1, I-309, inflammatory cytokine I-309, P500, SCYA1Small-inducible cytokine A1, SISe, small inducible cytokine A1 (I-309, homologous to mouse Tca-3), T lymphocyte-secreted protein I-309, TCA3
Mouse recombinant CCL1/I-309/TCA-3
0.1 mL
Chemokines and Cytokines
6346
Store at 4C in the dark.
IgG2