missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL11/Eotaxin Antibody (43915) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3201JF635
This item is not returnable.
View return policy
Description
CCL11/Eotaxin Monoclonal antibody specifically detects CCL11/Eotaxin in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| CCL11/Eotaxin | |
| Monoclonal | |
| Janelia Fluor 635 | |
| C-C motif chemokine 11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, eotaxin, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin) | |
| E. coli-derived recombinant human CCL11/Eotaxin | |
| 0.5 mL | |
| Asthma, Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology | |
| 6356 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 43915 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction