missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL12/MCP-5 Antibody (RM0169-8J25), Alexa Fluor™ 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12089AF594
This item is not returnable.
View return policy
Description
CCL12/MCP-5 Monoclonal specifically detects CCL12/MCP-5 in Mouse samples. It is validated for Western Blot.
Specifications
| CCL12/MCP-5 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Ccl12 | |
| Mouse recombinant CCL12/MCP-5 | |
| 0.1 mL | |
| 20293 | |
| Store at 4°C in the dark. |
| Western Blot | |
| RM0169-8J25 | |
| Western Blot | |
| C-C motif chemokine 12, chemokine (C-C motif) ligand 12, MCP-5, Scya12 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Mouse | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction