missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL14/HCC-1/HCC-3 Antibody (007), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89519F
This item is not returnable.
View return policy
Description
CCL14/HCC-1/HCC-3 Monoclonal antibody specifically detects CCL14/HCC-1/HCC-3 in Human samples. It is validated for ELISA, ELISA
Specifications
| CCL14/HCC-1/HCC-3 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 007 | |
| ELISA, Sandwich ELISA | |
| C-C motif chemokine 14, chemokine (C-C motif) ligand 14, Chemokine CC-1/CC-3, chemokine CC-3, CKB1, FLJ16015, HCC-1, HCC-1(1-74), HCC-1/HCC-3, HCC-3, hemofiltrate CC chemokine 1, MCIF, member 14, NCC2CC-1, NCC-2CKb1, new CC chemokine 2, SCYA14CC-3, Small-inducible cytokine A14 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL14/HCC-1/HCC-3 (Q16627-1; Thr20-Asn93). | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines | |
| 6358 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction