missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL15/MIP-1 delta Antibody (001), DyLight 550, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89703R
This item is not returnable.
View return policy
Description
CCL15/MIP-1 delta Monoclonal antibody specifically detects CCL15/MIP-1 delta in Human samples. It is validated for ELISA
Specifications
| CCL15/MIP-1 delta | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 001 | |
| ELISA | |
| chemokine (C-C motif) ligand 15, Chemokine CC-2, HCC-2Lkn-1, leukotactin 1, Leukotactin-1, LKN1, LKN-1, Macrophage inflammatory protein 5, MIP-1 delta, MIP5, MIP-5MIP-1d, Mrp-2b, NCC-3MIP-1D, NCC3MRP-2B, new CC chemokine 3, SCYA15member 15, Small-inducible cytokine A15 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL15/MIP-1 delta (Q16663-1; Gln22-Ile113). | |
| 0.1 mL | |
| Biologically Active Proteins, Chemokines and Cytokines, Protein Kinase | |
| 6359 | |
| Store at 4°C in the dark. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto