Learn More
CCL16/HCC-4/LEC Antibody (013), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP2-89529CL1
Product Code. 30153140
  • € 1020.00 / 0.10mL
Description
CCL16/HCC-4/LEC Monoclonal antibody specifically detects CCL16/HCC-4/LEC in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCL16/HCC-4/LEC
Monoclonal
CoraFluor 1
C-C motif chemokine 16, chemokine (C-C motif) ligand 16, Chemokine CC-4, Chemokine LEC, CKb12, HCC-4Liver-expressed chemokine, LCC-1Lymphocyte and monocyte chemoattractant, LEC, liver CC chemokine-1, LMCSmall-inducible cytokine A16, monotactin-1, MTN-1, NCC4IL-10-inducible chemokine, NCC-4ILINCK, new CC chemokine 4, SCYA16MGC117051, SCYL4, small inducible cytokine subfamily A (Cys-Cys), member 16
This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL16/HCC-4/LEC (Accession#: NP_004581.1; Gln24-Gln120).
0.1 mL
Biologically Active Proteins, Chemokines and Cytokines
6360
Store at 4°C in the dark. Do not freeze.
IgG
ELISA
013
PBS
Rabbit
Protein A purified