missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL17/TARC Antibody (54015), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3641V
This item is not returnable.
View return policy
Description
CCL17/TARC Monoclonal antibody specifically detects CCL17/TARC in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| CCL17/TARC | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 54015 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARCA-152E5.3, Thymus and activation-regulated chemokine | |
| E. coli-derived recombinant human CCL17/TARC, Ala24-Ser94, Accession # Q92583.1 | |
| 0.5 mL | |
| Biologically Active Proteins, Chemokines and Cytokines | |
| 6361 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction