missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL18/PARC Antibody (016), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89535DL594
This item is not returnable.
View return policy
Description
CCL18/PARC Monoclonal antibody specifically detects CCL18/PARC in Human samples. It is validated for ELISA
Specifications
| CCL18/PARC | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 016 | |
| ELISA | |
| Alternative macrophage activation-associated CC chemokine 1, AMAC1CC chemokine ligand 18, AMAC-1Small-inducible cytokine A18, chemokine (C-C motif) ligand 18 (pulmonary and activation-regulated), CKb7, DC-CK1CC chemokine PARC, DCCK1Macrophage inflammatory protein 4, Dendritic cell chemokine 1, MIP4, MIP-4C-C motif chemokine 18, PARCPulmonary and activation-regulated chemokine, SCYA18chemokine (C-C), dendritic, small inducible cytokine A18, small inducible cytokine subfamily A (Cys-Cys), member 18, pulmonary andactivation-regulated | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL18/PARC (NP_002979.1; Ala21-Ala89). | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer | |
| 6362 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction