missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL19 Mouse anti-Human, PE, Clone: T50-867, BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566523
This item is not returnable.
View return policy
Description
CCL19 Monoclonal antibody specifically detects CCL19 in Human samples. It is validated for Flow Cytometry.
Specifications
| CCL19 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry | |
| T50-867 | |
| PE | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction