missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL2/JE/MCP-1 Antibody (017), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90551CL1
This item is not returnable.
View return policy
Description
CCL2/JE/MCP-1 Monoclonal antibody specifically detects CCL2/JE/MCP-1 in Mouse samples. It is validated for ELISA,Sandwich ELISA Capture
Specifications
| CCL2/JE/MCP-1 | |
| Monoclonal | |
| CoraFluor 1 | |
| C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, Mcp-1, SCYA2, SMC-CF | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CCL2/JE/MCP-1 (Accession#: NP_035463.1; Met1-Asn148). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| ELISA, Sandwich ELISA Capture | |
| 017 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 6347 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction