missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL2/JE/MCP-1 Antibody (24822), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB279H
This item is not returnable.
View return policy
Description
CCL2/JE/MCP-1 Monoclonal antibody specifically detects CCL2/JE/MCP-1 in Human samples. It is validated for Western Blot, Neutralization
Specifications
| CCL2/JE/MCP-1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6347 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Neutralization | |
| 24822 | |
| Western Blot, Neutralization | |
| C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, Mcp-1, SCYA2, SMC-CF | |
| E. coli-derived recombinant human CCL2/JE/MCP-1 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction