missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL2, Mouse anti-Human, Clone: 2E2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00006347-M08.50ug
This item is not returnable.
View return policy
Description
Sequence: MKVSAALLCLLLIAATFIPQGLAQPDAINAPVTCCYNFTNRKISVQRLASYRRITSSKCPKEAVIFKTIVAKEICADPKQKWVQDSMDHLDKQTQTPKTSpecifications
| CCL2 | |
| Monoclonal | |
| Unconjugated | |
| chemokine (C-C motif) ligand 2 | |
| GDCF-2/HC11/HSMCR30/MCAF/MCP-1/MCP1/MGC9434/SCYA2/SMC-CF | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 2E2 | |
| In 1x PBS, pH 7.4 | |
| BC009716 | |
| CCL2 | |
| CCL2 (AAH09716, 1 a.a. ∼ 99 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6347 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction