missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL20/MIP-3 alpha Antibody (114906), DyLight 405, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB760E
This item is not returnable.
View return policy
Description
CCL20/MIP-3 alpha Monoclonal antibody specifically detects CCL20/MIP-3 alpha in Mouse samples. It is validated for Western Blot
Specifications
| CCL20/MIP-3 alpha | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| 114906 | |
| Western Blot | |
| beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38 | |
| E. coli-derived recombinant mouse CCL20/MIP-3 alpha , Ala28-Met97, Accession # O89093 | |
| 0.1 mL | |
| Biologically Active Proteins, Chemokines and Cytokines | |
| 6364 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction