Learn More
CCL21/6Ckine Antibody (59106), Alexa Fluor™ 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB457T
Product Code. 30064585
  • € 642.00 / 0.10mL
Description
CCL21/6Ckine Monoclonal antibody specifically detects CCL21/6Ckine in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCL21/6Ckine
Monoclonal
Alexa Fluor 594
50mM Sodium Borate
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Mouse
Purified
Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
59106
Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, CyTOF-ready
6CkineSmall-inducible cytokine A21, Beta-chemokine exodus-2, chemokine (C-C motif) ligand 21, CKb9, member 21, SCYA21MGC34555, secondary lymphoid tissue chemokine, SLCSecondary lymphoid-tissue chemokine
E. coli-derived recombinant mouse CCL21/6Ckine, Ser24-Gly133, Accession # P84444
0.1 mL
Biologically Active Proteins, Cancer, Cell Cycle and Replication, Chemokines and Cytokines
6366
Store at 4C in the dark.
IgG2b