Learn More
CCL22/MDC Antibody (57203), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB3361C
Product Code. 30039834
  • ONLINE EXCLUSIVE PRICE
    € 896.00
    € 847.35 / 0.50mL
    Save € 48.65
    5% Off
Description
CCL22/MDC Monoclonal antibody specifically detects CCL22/MDC in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCL22/MDC
Monoclonal
PerCP
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, CyTOF
57203
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1
E. coli-derived recombinant human CCL22/MDC, Gly25-Gln93, Accession # O00626.1
0.5 mL
Biologically Active Proteins
6367
Store at 4C in the dark.
IgG2b