missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL24/Eotaxin-2/MPIF-2 Antibody (61016.11), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-10370H
This item is not returnable.
View return policy
Description
CCL24/Eotaxin-2/MPIF-2 Monoclonal specifically detects CCL24/Eotaxin-2/MPIF-2 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| CCL24/Eotaxin-2/MPIF-2 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| CCL24 | |
| Purified recombinant human eotaxin 2, expressed in E. coli as immunogen. | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 6369 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 61016.11 | |
| Western Blot, ELISA | |
| chemokine (C-C motif) ligand 24, Ckb-6, Eosinophil chemotactic protein 2, Eotaxin-2, member 24, MPIF2Myeloid progenitor inhibitory factor 2, MPIF-2Small-inducible cytokine A24, SCYA24CK-beta-6 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| When used as a capture in human eotaxin 2 sandwich ELISAs, this exhibits less than 0.03 % cross-reactivity with recombinant human eotaxin, MIP1 alpha, and MCP3. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction