missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL25/TECK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 589.00
Specifications
| Antigen | CCL25/TECK |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18662738
|
Novus Biologicals
NBP2-49199-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603998
|
Novus Biologicals
NBP2-49199 |
0.1 mL |
€ 589.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL25/TECK Polyclonal antibody specifically detects CCL25/TECK in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CCL25/TECK | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Chemokines and Cytokines, Signal Transduction | |
| PBS (pH 7.2), 40% Glycerol | |
| 6370 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| C-C motif chemokine 25, chemokine (C-C motif) ligand 25, Chemokine TECK, Ck beta-15, Ckb15, SCYA25TECKvar, small inducible cytokine subfamily A (Cys-Cys), member 25, Small-inducible cytokine A25, TECKMGC150327, thymus expressed chemokine, Thymus-expressed chemokine | |
| This antibody was developed against a recombinant protein corresponding to amino acids: HTQGVFEDCCLAYHYPIGWAVL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title