missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL3/MIP-1 alpha Antibody (39624), Alexa Fluor™ 700, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB450N
This item is not returnable.
View return policy
Description
CCL3/MIP-1 alpha Monoclonal antibody specifically detects CCL3/MIP-1 alpha in Mouse samples. It is validated for Western Blot
Specifications
| CCL3/MIP-1 alpha | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| 39624 | |
| Western Blot | |
| C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78alpha, Mip1a, MIP-1alpha, MIP1-alpha, Scya3 | |
| E. coli-derived recombinant mouse CCL3/MIP-1 alpha , Ala24-Ala92, Accession # P10855 | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
| 6348 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction