missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL3, Mouse anti-Human, Clone: 2E9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00006348-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: SLAADTPTACCFSYTSRQIPQNFIADYFETSSQCSKPGVIFLTKRSRQVCADPSEEWVQKYVSDLELSASpecifications
| CCL3 | |
| Monoclonal | |
| Unconjugated | |
| chemokine (C-C motif) ligand 3 | |
| G0S19-1/LD78ALPHA/MIP-1-alpha/MIP1A/SCYA3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 2E9 | |
| In 1x PBS, pH 7.4 | |
| NM_002983 | |
| CCL3 | |
| CCL3 (NP_002974, 24 a.a. ∼ 92 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6348 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction