missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL6/C10 Antibody (262016), Alexa Fluor™ 700, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB487N
This item is not returnable.
View return policy
Description
CCL6/C10 Monoclonal antibody specifically detects CCL6/C10 in Mouse samples. It is validated for Western Blot, Neutralization, Flow Cytometry, CyTOF
Specifications
| CCL6/C10 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Neutralization, Flow Cytometry, CyTOF | |
| 262016 | |
| Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| c10,C-C motif chemokine 6,MRP,MRP-1,Scy,Scya6 | |
| E. coli-derived recombinant mouse CCL6/C10, Gly22-Ala116, Accession # P27784 | |
| 0.5 mL | |
| Chemokines and Cytokines | |
| 20305 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction