missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL6/C10 Antibody (RM0183-7J13), Alexa Fluor™ 700, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12420AF700
This item is not returnable.
View return policy
Description
CCL6/C10 Monoclonal specifically detects CCL6/C10 in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| CCL6/C10 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Ccl6 | |
| Mouse recombinant CCL6/C10 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry | |
| RM0183-7J13 | |
| Western Blot, Flow Cytometry | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 20305 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction