missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL6/C10 Antibody (RM0183-7J13), Janelia Fluor™ 549, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12420JF549
This item is not returnable.
View return policy
Description
CCL6/C10 Monoclonal specifically detects CCL6/C10 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CCL6/C10 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Rat | |
| Protein A or G purified | |
| Chemokines and Cytokines | |
| 20305 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Western Blot, Flow Cytometry | |
| RM0183-7J13 | |
| Western Blot, Flow Cytometry | |
| Ccl6 | |
| Mouse recombinant CCL6/C10 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction