missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL6/C10 Antibody (RM0183-7J13), PE/Atto594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12420PEATT594
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
CCL6/C10 Monoclonal specifically detects CCL6/C10 in Mouse samples. It is validated for Flow Cytometry.
Tekniske data
| CCL6/C10 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| Rat | |
| Protein A or G purified | |
| Chemokines and Cytokines | |
| 20305.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| RM0183-7J13 | |
| Flow Cytometry | |
| Ccl6 | |
| Mouse recombinant CCL6/C10 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG2 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion