missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL7/MCP-3/MARC Antibody (063), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90018AF405
This item is not returnable.
View return policy
Description
CCL7/MCP-3/MARC Monoclonal specifically detects CCL7/MCP-3/MARC in Human samples. It is validated for ELISA.
Specifications
| CCL7/MCP-3/MARC | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 63 | |
| ELISA | |
| C-C motif chemokine 7, chemokine (C-C motif) ligand 7, FIC, MARC, MCP3Monocyte chemotactic protein 3, MCP-3Small-inducible cytokine A7, MGC138465, Monocyte chemoattractant protein 3, NC28SCYA6, SCYA7MGC138463, small inducible cytokine A7 (monocyte chemotactic protein 3) | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL7/MCP-3/MARC (NP_006264.2; Gln24-Leu99). | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, Chemokines and Cytokines | |
| 6354 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction