Learn More
CCL9 Antibody (RM0185-28B3), Alexa Fluor™ 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12408AF594
This item is not returnable.
View return policy
Description
CCL9 Monoclonal specifically detects CCL9 in Mouse samples. It is validated for Western Blot.
Specifications
| CCL9 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CCL9 | |
| Mouse recombinant CCL9 | |
| 0.1 mL | |
| 20308 | |
| Store at 4°C in the dark. |
| Western Blot | |
| RM0185-28B3 | |
| Western Blot | |
| CCF18, Ccl9 chemokine (C-C motif) ligand 9, MRP-2, Scya10, Scya9 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Mouse | |
| IgG2 |