missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CCMI

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Positive allosteric modulator of α7 neuronal nicotinic acetylcholine receptors (nAChR). Evokes positive modulation of acetylcholine (ACh)-induced EC5 currents (EC50 = 0.7 μM). Exhibits cognitive-enhancing properties in rodent models; displays no cytotoxic effects in PC12 cells or rat primary cortical neurons.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 388.25 |
| Purity | >98% |
| Molecular Weight (g/mol) | 388.248 |
| Chemical Name or Material | CCMI |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction