Learn More
CCNB1 Mouse anti-Human, Clone: SPM619, Abnova™
Mouse monoclonal antibody raised against full length recombinant human CCNB1.
Brand: Abnova MAB13180.100ug
Description
The protein encoded by this gene is a regulatory protein involved in mitosis. The gene product complexes with p34(cdc2) to form the maturation-promoting factor (MPF). Two alternative transcripts have been found, a constitutively expressed transcript and a cell cycle-regulated transcript, that is expressed predominantly during G2/M phase. The different transcripts result from the use of alternate transcription initiation sites. [provided by RefSeq]
Specifications
| CCNB1 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| CCNB | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 891 | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| SPM619 | |
| Mouse monoclonal antibody raised against full length recombinant human CCNB1. | |
| In 10mM PBS. | |
| CCNB1 | |
| Recombinant protein corresponding to full length human CCNB1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.