missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCNB1IP1 Antibody (OTI4H3), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72106F
This item is not returnable.
View return policy
Description
CCNB1IP1 Monoclonal specifically detects CCNB1IP1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
| CCNB1IP1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| CCNB1IP1 | |
| Full length recombinant protein of human CCNB1IP1 (NP_878269) produced in E.coli. | |
| 0.1 mL | |
| 57820.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry | |
| OTI4H3 | |
| Flow Cytometry, Immunohistochemistry | |
| chromosome 14 open reading frame 18, cyclin B1 interacting protein 1, cyclin B1 interacting protein 1, E3 ubiquitin protein ligase, E3 ubiquitin-protein ligase CCNB1IP1, EC 6.3.2.-, enhancer of invasion 10, HEI10C14orf18Cyclin-B1-interacting protein 1, Human enhancer of invasion 10 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction