Learn More
CCND2 Mouse anti-Human, Clone: OTI7G9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF806254
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cyclin D2 is a regulatory component of the cyclin D2-CDK4 (DC) complex that phosphorylates and inhibits members of the retinoblastoma (RB) protein family including RB1 and regulates the cell-cycle during G1/S transition. Phosphorylation of RB1 allows dissociation of the transcription factor E2F from the RB/E2F complex and the subsequent transcription of E2F target genes which are responsible for the progression through the G1 phase. Hypophosphorylates RB1 in early G1 phase. Cyclin D-CDK4 complexes are major integrators of various mitogenenic and antimitogenic signals. Also substrate for SMAD3, phosphorylating SMAD3 in a cell-cycle-dependent manner and repressing its transcriptional activity. Component of the ternary complex, cyclin D2/CDK4/CDKN1B, required for nuclear translocation and activity of the cyclin D-CDK4 complex.Specifications
| CCND2 | |
| Monoclonal | |
| Unconjugated | |
| CCND2 | |
| KIAK0002, MPPH3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 894 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI7G9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P30279 | |
| CCND2 | |
| Full length human recombit protein of human CCND2 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.