missing translation for 'onlineSavingsMsg'
Learn More

CCNE2 Rabbit, anti-Human, Polyclonal, Proteintech

Product Code. 18070306 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18070306 150 μL 150µL
18074735 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18070306 Supplier Proteintech Supplier No. 307681AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The CCNE2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CCNE2. This antibody recognizes human, mouse, rat antigen. The CCNE2 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigen CCNE2
Applications Western Blot, ELISA
Concentration 400 μg/mL
Gene Symbols CCNE2, CYCE2, cyclin E2, G1/S specific cyclin E2
Host Species Rabbit
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 9134
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.