missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR1 Antibody (643854), DyLight 650, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB5986W
This item is not returnable.
View return policy
Description
CCR1 Monoclonal antibody specifically detects CCR1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| CCR1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 643854 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| C-C chemokine receptor type 1, CC-CKR-1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
| E. coli-derived recombinant mouse CCR1, Extracellular regions (aa 1-34, aa 92-107, aa 172-192, aa 265-281), Accession # P51675 | |
| 0.1 mL | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1230 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction