missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR10/GPR2 Antibody (314305), PE/Cy5.5, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB3478PECY55
This item is not returnable.
View return policy
Description
CCR10/GPR2 Monoclonal antibody specifically detects CCR10/GPR2 in Human samples. It is validated for Flow Cytometry
Specifications
| CCR10/GPR2 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 314305 | |
| Flow Cytometry | |
| CC chemokine receptor 10, C-C chemokine receptor type 10, C-C CKR-10, CC-CKR-10, CCR-10, chemokine (C-C motif) receptor 10, G protein-coupled receptor 2, GPR2G-protein coupled receptor 2 | |
| Y3 rat myeloid cell line transfected with human CCR10, Gln8-Asn362, Accession # P46092 | |
| 0.1 mL | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 2826 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction