missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCR11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575961
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 96% (by SDS-PAGE).
Receptors for the C-C chemokine family include CCR-1, CCR-2A, CCR-3, CCR-4, CCR-5 and the Duffy blood group antigen. The C-C receptors are important in the function of T cell chemotaxis and migration of phagocytic cells to sites of inflammation. CCR11 in particular is expressed in the heart, lung, and small intestine. It binds members of the monocyte chemoattractant family including MCP-1, MCP-2, and MCP-4.
Specifications
| CCR11 | |
| Polyclonal | |
| Unconjugated | |
| ACKR4 | |
| A630091E18Rik; Ackr4; Atypical chemokine receptor 4; C-C chemokine receptor type 11; CC chemokine receptor-like 1; C-C CKR-11; CCBP2; CC-CKR-11; CCR10; CCR11; CCR-11; Ccrl1; CCX CKR; CCX-CKR; CCX-CKR1; chemocentryx chemokine receptor; chemokine (C-C motif) receptor-like 1; chemokine (C-C) receptor-like 1; chemokine receptor CCR11; chemokine, cc motif, receptor-like protein 1; CKR-11; Cmkbrl1; orphan seven-transmembrane receptor, chemokine related; PPR1; VSHK1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 252837, 51554, 685243 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q924I3, Q9NPB9 | |
| ACKR4 | |
| Synthetic peptide corresponding to amino acids 298-342 of Human CCR11. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction