missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR2 Antibody (3G7), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-35334AF594
This item is not returnable.
View return policy
Description
CCR2 Monoclonal specifically detects CCR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CCR2 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CCR2 | |
| A synthetic peptide made to an N-terminal portion of the human CD63 protein (between residues 1-100) [UniProt P41597] | |
| Primary | |
| Human | |
| IgG2b λ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 3G7 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2CKR2B, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
| Mouse | |
| 0.1 mL | |
| 729230 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction