missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR2 Antibody (3G7), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-35334FR
This item is not returnable.
View return policy
Description
CCR2 Monoclonal specifically detects CCR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CCR2 | |
| Monoclonal | |
| DyLight 680 | |
| C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2CKR2B, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Immunogen's sequence similarity with other species: Mouse (57%), Rabbit (57%). | |
| Store at 4C in the dark. | |
| IgG2b λ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 3G7 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CCR2 | |
| A synthetic peptide made to an N-terminal portion of the human CD63 protein (between residues 1-100) [UniProt P41597] | |
| 0.1 mL | |
| Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 729230 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction