missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR2 Antibody (890231R), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8368RMFV500
This item is not returnable.
View return policy
Description
CCR2 Monoclonal antibody specifically detects CCR2 in Rat samples. It is validated for Flow Cytometry, CyTOF
Specifications
| CCR2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 729230 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 890231R | |
| Flow Cytometry, CyTOF-ready | |
| C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2CKR2B, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
| NS0 mouse myeloma cell line transfected with rat CCR2, Accession # O55193 | |
| RUO | |
| Primary | |
| Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction