missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR3 Antibody (61828) [Biotin], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB155B
This item is not returnable.
View return policy
Description
CCR3 Monoclonal antibody specifically detects CCR3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF
Specifications
| CCR3 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF | |
| 61828 | |
| Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready | |
| b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR-3, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841 | |
| Y3 rat myeloid cell line transfected with human CCR3, Accession # P51677 | |
| 0.1 mL | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1232 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction