missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR4 Antibody (MM0064-9G12), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12129AF350
This item is not returnable.
View return policy
Description
CCR4 Monoclonal specifically detects CCR4 in Human samples. It is validated for Flow Cytometry.
Specifications
| CCR4 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CCR4 | |
| Human CCR4 transfected 293 cells | |
| Primary | |
| Detects human CCR4, not other CCRs | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| MM0064-9G12 | |
| Flow Cytometry, Immunohistochemistry-Frozen | |
| C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5 | |
| Mouse | |
| 0.1 mL | |
| 1233 | |
| Human | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction