missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR4 Antibody (MM0064-9G12), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12129PCP
This item is not returnable.
View return policy
Description
CCR4 Monoclonal specifically detects CCR4 in Human samples. It is validated for Flow Cytometry.
Specifications
| CCR4 | |
| Monoclonal | |
| PerCP | |
| P51679 | |
| CCR4 | |
| Human CCR4 transfected 293 cells | |
| 0.1 mL | |
| Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1233 | |
| Human | |
| Purified |
| Flow Cytometry | |
| MM0064-9G12 | |
| Flow Cytometry | |
| C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects human CCR4, not other CCRs | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction