missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR5 Antibody (225307), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB18041MFV610
This item is not returnable.
View return policy
Description
CCR5 Monoclonal antibody specifically detects CCR5 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| CCR5 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Alzheimers Research, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction | |
| 1234 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 225307 | |
| Flow Cytometry, CyTOF-ready | |
| C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 | |
| C6 rat glioma cell line transfected with mouse CCR5, Accession # P51682 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction