missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR6 Antibody (18B9E6), DyLight 405, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-25224V
This item is not returnable.
View return policy
Description
CCR6 Monoclonal specifically detects CCR6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CCR6 | |
| Monoclonal | |
| DyLight 405 | |
| BN-1, CC-CKR-6, CCR-6, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29C-C CKR-6, GPRCY4DRY-6, lymphocyte, 22 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| This product reacts with Mouse. | |
| Store at 4C in the dark. | |
| IgG2a κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 18B9E6 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CCR6 | |
| A recombinant protein consisting of the external domains of mouse CD196 (CCR6) was used as the immunogen for the antibody. | |
| 0.1 mL | |
| Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1235 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction