missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR7 Antibody (4B12) [PerCP], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB3477C
This item is not returnable.
View return policy
Description
CCR7 Monoclonal antibody specifically detects CCR7 in Mouse samples. It is validated for Flow Cytometry, Neutralization, Immunocytochemistry, CyTOF-reported
Specifications
| CCR7 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Neutralization, Immunocytochemistry, CyTOF | |
| 4B12 | |
| Flow Cytometry, Neutralization, Immunocytochemistry, CyTOF-reported | |
| BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR-7, CD197, CD197 antigen, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7chemokine (C-C) receptor 7, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor | |
| RBC-2H3 cells expressing mouse CCR7, Accession # NP_031745 | |
| 0.1 mL | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1236 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction