missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR7 Mouse anti-Human, Clone:4B7B8, Abnova™
Mouse monoclonal antibody raised against recombinant human CCR7.
Brand: Abnova MAB17599.100ug
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the G protein-coupled receptor family. This receptor was identified as a gene induced by the Epstein-Barr virus (EBV), and is thought to be a mediator of EBV effects on B lymphocytes. This receptor is expressed in various lymphoid tissues and activates B and T lymphocytes. It has been shown to control the migration of memory T cells to inflamed tissues, as well as stimulate dendritic cell maturation. The chemokine (C-C motif) ligand 19 (CCL19/ECL) has been reported to be a specific ligand of this receptor. [provided by RefSeq]Specifications
| CCR7 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunocytochemistry Flow Cytometry (1:200-1:400) Immunohistochemistry The optimal working dilution should be determined by the end user. |
|
| CCR7 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 4B7B8 | |
| chemokine (C-C motif) receptor 7 | |
| In PBS (0.05% sodium azide) | |
| BLR2, CD197, CDw197, CMKBR7, EBI1 | |
| Recombinant protein corresponding to amino acid of human CCR7 from E. coli. | |
| 1236 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction