missing translation for 'onlineSavingsMsg'
Learn More

CCR8 Antibody (6B12), Novus Biologicals™

Human Monoclonal Antibody

€ 343.00 - € 533.00

Specifications

Antigen CCR8
Clone 6B12
Dilution Flow Cytometry
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043440
View Documents
Novus Biologicals
NBP3-26687-50ul
50 μL
€ 343.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043164
View Documents
Novus Biologicals
NBP3-26687-100ul
100 μL
€ 533.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CCR8 Monoclonal antibody specifically detects CCR8 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CCR8
Flow Cytometry
Monoclonal
Purified
RUO
Human
CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR-8, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCY6, GPR-CY6, TER1CC chemokine receptor CHEMR1
Recombinant Human CCR8 protein [UniProt P51685]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
6B12
Flow Cytometry
Unconjugated
Human
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites
50% Glycerol, 0.01M PBS, PH 7.4
1237
IgG1
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.